Skip to Content
Merck
All Photos(1)

Key Documents

X3254

Sigma-Aldrich

Xylanase 1, thermostable

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

Xylanase , thermostable

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352204

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

liquid

specific activity

≥40 units/mg protein

mol wt

45 kDa

concentration

≥20 mg protein/mL (Bradford)

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Xylanase is a member of a family of glycoside hydrolases responsible for the breakdown of xylan in plants by cleaving the β 1,4 backbone.

Unit Definition

One unit will produce 1 μmole of reducing sugar (measured as xylose) from xylan per minute at pH 5.8 at 70 °C.

Physical form

Supplied as a solution in 50 mM Tris-HCl, pH 7.5, 100 mM NaCl, and 25% glycerol.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tony Collins et al.
FEMS microbiology reviews, 29(1), 3-23 (2005-01-18)
Xylanases are hydrolytic enzymes which randomly cleave the beta 1,4 backbone of the complex plant cell wall polysaccharide xylan. Diverse forms of these enzymes exist, displaying varying folds, mechanisms of action, substrate specificities, hydrolytic activities (yields, rates and products) and
Ming-Qi Liu et al.
Protein expression and purification, 88(1), 85-92 (2012-12-19)
The xylan binding domain (XBD) and linker sequences (LS) from thermostable and thermophilic Thermomonospora fusca xylanase A (TfxA) was fused to the carboxyl-terminus of a family 11 hybrid xylanase ATx. The constructed chimera (ATxX) was successfully expressed in Pichia pastoris
Dong-Ju Park et al.
Journal of microbiology and biotechnology, 23(3), 397-404 (2013-03-07)
Paenibacillus xylanilyticus KJ-03 was isolated from soil samples obtained from a field with Amorphophallus konjac plants. A gene encoding xylanase was isolated from KJ-03 and cloned using a fosmid library. The xynA gene encodes xylanase; it consists of 1,035 bp
Digvijay Verma et al.
PloS one, 8(1), e52459-e52459 (2013-02-06)
The alkalistable and thermostable xylanases are in high demand for pulp bleaching in paper industry and generating xylooligosaccharides by hydrolyzing xylan component of agro-residues. The compost-soil samples, one of the hot environments, are expected to be a rich source of
Rou Min et al.
Wei sheng wu xue bao = Acta microbiologica Sinica, 53(4), 346-352 (2013-07-19)
To reveal the correlation between thermostability of xylanase EvXyn11(TS) and its N-terminal disulfide bridge, an EvXyn11(TS)-encoding gene (Syxyn11) was synthesized and subjected to site-directed mutagenesis. Multiple homology alignment of protein primary structures between the EvXyn11(TS) and several GH family 11

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service