Skip to Content
Merck
All Photos(1)

Key Documents

T5772

Sigma-Aldrich

Thrombin from rat plasma

lyophilized powder, ≥1,000 units/mg protein (biuret)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

rat plasma

Quality Level

form

lyophilized powder

specific activity

≥1,000 units/mg protein (biuret)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... F2(29251)
rat ... F2(29251)

General description

Thrombin is the final coagulation protease in regard to hemostasis, promoting both procoagulant and anticoagulant effects.

Application

Thrombin is used for site specific cleavage of recombinant fusion proteins containing an accessible thrombin recognition site for removal of affinity tags.
Thrombin is used for site specific cleavage of recombinant fusion proteins containing an accessible thrombin recognition site for removal of affinity tags. Thrombin has been used in a study to investigate activation of equine platelet-rich plasma. [1]

Biochem/physiol Actions

Serine protease that selectively cleaves Arg-Gly bonds in fibrinogen to form fibrin and fibrinopeptides A and B.

Unit Definition

Activity is expressed in NIH units obtained by direct comparison to a NIH Thrombin Reference Standard, Lot K.

Physical form

Lyophilized from saline sodium citrate buffer, pH 6.5

Analysis Note

The NIH assay procedure uses 0.2 ml diluted plasma (1:1 with saline) as a substrate and 0.1 ml of thrombin sample (stabilized in a 1% buffered albumin solution) based on a modification of the method of Biggs. Only clotting times in the range of 15-25 seconds are used for determining thrombin concentrations.

Other Notes

View more information on thrombin at www.sigma-aldrich.com/enzymeexplorer.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaojun Chen et al.
Cell death discovery, 8(1), 189-189 (2022-04-12)
Spinal cord injury (SCI) will result in the significant elevation of thrombin production at lesion site via either breakage of blood-spinal cord barrier or upregulated expression within nerve cells. Thrombin-induced activation of the protease activated receptors (PARs) evokes various pathological
Jamie A Textor et al.
Veterinary surgery : VS, 41(7), 784-794 (2012-06-30)
To investigate and compare clinically relevant Platelet-rich plasma (PRP) activation methods. Experimental. PRP was prepared from 6 equine subjects. Activation of the PRP was performed by 4 methods (autologous thrombin, bovine thrombin, calcium chloride (CaCl(2) ), or freeze-thaw). The resultant
Ting Yang et al.
Journal of neuroinflammation, 19(1), 120-120 (2022-05-28)
The danger-associated molecular patterns (DAMPs) are critical contributors to the progressive neuropathology and thereafter affect the functional outcomes following spinal cord injury (SCI). Up to now, the regulatory mechanisms on their inducible production from the living cells remain elusive, aside
Dilyara Cheranova et al.
Journal of visualized experiments : JoVE, (72)(72), doi:10-doi:10 (2013-02-22)
The characterization of gene expression in cells via measurement of mRNA levels is a useful tool in determining how the transcriptional machinery of the cell is affected by external signals (e.g. drug treatment), or how cells differ between a healthy
Response.
Sarah T Garber et al.
Journal of neurosurgery, 118(2), 485-485 (2013-03-16)

Articles

Thrombin Factor IIa is an endolytic serine protease that selectively cleaves the Arg--Gly bonds of fibrinogen to form fibrin and release fibrinopeptides A and B.

Thrombin Factor IIa is an endolytic serine protease that selectively cleaves the Arg--Gly bonds of fibrinogen to form fibrin and release fibrinopeptides A and B.

Thrombin Factor IIa is an endolytic serine protease that selectively cleaves the Arg--Gly bonds of fibrinogen to form fibrin and release fibrinopeptides A and B.

Thrombin Factor IIa is an endolytic serine protease that selectively cleaves the Arg--Gly bonds of fibrinogen to form fibrin and release fibrinopeptides A and B.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service