Skip to Content
Merck
All Photos(1)

Key Documents

SRP6594

Sigma-Aldrich

ErbB4/HER4 HIS tag human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

MGC138404, p180erbB4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

tag

His tagged

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

5.8 kDa

packaging

pkg of 5 μg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ERBB4(2066)

General description

Akt3 or Protein Kinase B γ (PKB) is a serine/threonine kinase that is a member of the Akt family. In mammals, the Akt family comprises of three highly homologous members known as Akt1 (PKB), Akt2 (PKB), and Akt 3. Akt 3 like the other family members is activated in cells exposed to diverse stimuli such as hormones, growth factors, and extracellular matrix components. The activation mechanism remains to be fully characterized but occurs downstream of phosphoinositide 3-kinase (PI3K). PI3K generates phosphatidylinositol-3,4,5-trisphosphate (PIP3), a lipid second messenger essential for the translocation of Akt family members to the plasma membrane where they are phosphorylated and activated by phosphoinositide-dependent kinase-1 (PDK-1) and phosphoinositide-dependent kinase-2 (PDK-2 possibly ILK). Akt 3 like the other family members phosphorylates and regulates the function of many cellular proteins involved in processes that include cellular metabolism, survival/apoptosis, and proliferation. Recent evidence indicates that Akt 3 is frequently overexpressed in many types of human cancers including breast and prostate. Although the mechanisms have not yet been fully characterized, increased expression and activation of Akt is believed to promote cell proliferation and survival thereby contributing to cancer progression.

Physical form

Sterile filtered white lyophilized (freeze-dried) powder. The protein was lyophilized from 50mM Sodium Chloride solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service