Skip to Content
Merck
All Photos(1)

Key Documents

SRP6503

Sigma-Aldrich

Notch-1 from mouse

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

Neurogenic Locus Notch Homolog Protein 1, Notch A, mT14, p300

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt ~85.0 kDa

packaging

pkg of 25 μg

impurities

<0.1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... NOTCH1(18128)

General description

Notch signaling pathway regulates many different cell fate decisions in both vertebrate and invertebrate species. There are 5 canonical Notch ligands in mammals: Jagged-1, Jagged-2, DLL1, DLL3 and DLL4. These can bind to the four Notch receptors Notch 1-4. It is important for pattern formation during development such as neurogenesis, angiogenesis or myogenesis and regulates T cell development and stem cell maintenance. Notch signaling is also involved in cellular processes through-out adulthood. Signaling via Notch occurs between neighbouring cells and both the receptor and its ligands are transmembrane proteins.
The extracellular domain of mouse Notch-1 (aa 19-488) (12 epidermal growth factor-like (EGF) repeats)

Physical form

Lyophilized with PBS.

Reconstitution

Reconstitute with sterile water to 1 mg/mL.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service