Skip to Content
Merck
All Photos(1)

Key Documents

SRP6327

Sigma-Aldrich

CD274 from mouse

recombinant, expressed in E. coli, ≥85% (SDS-PAGE)

Synonym(s):

A530045L16Rik, B7h1, CD274 antigen, Pdcd1l1, Pdcd1lg1, Pdl1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

Assay

≥85% (SDS-PAGE)

form

liquid

mol wt

27.6 kDa (246 aa, 19-239 aa + His tag)

packaging

pkg of 100 μg

concentration

1 mg/mL

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... CD274(60533)

General description

Programmed cell death 1 ligand 1 (PD-L1), which is also known as cluster of differentiation (CD274) or B7 homolog 1 (B7-H1), is a member of the growing B7 family of immune molecules. PD-L1 is a cell surface immunoglobulin with two Ig-like domains within the extracellular region and a short cytoplasmic domain. The protein is highly expressed in the heart, skeletal muscle, placenta and lung and weakly expressed in the thymus, spleen, kidney and liver. PD-L1 is expressed on activated T-cells, B-cells, dendritic cells, keratinocytes and monocytes. Recombinant mouse CD274 protein, fused to His-tag at N-terminus, was expressed in E.coli and purified by using conventional chromatography.

Biochem/physiol Actions

Programmed cell death 1 ligand 1 (PD-L1) is involved in the regulation of cellular and humoral immune responses. PD-L1 is up-regulated on T- and B-cells, dendritic cells, keratinocytes and monocytes after lipopolysaccharide (LPS) and interferon-γ (IFNG) activation and up-regulated in B-cells activated by surface Ig cross-linking. PD-L1 is involved in the co-stimulatory signal, essential for T-cell proliferation and production of interleukin-10 (IL10) and IFNG, in an interleukin-2-dependent and a programmed cell death 1 (PDCD1)-independent manner.

Physical form

1 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) containing 0.15 M NaCl and 10% glycerol.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of B7 family molecules in hematologic malignancy.
Greaves P and Gribben JG
Blood, 121(5), 734-744 (2013)
Intrinsic and extrinsic control of expression of the immunoregulatory molecule PD-L1 in epithelial cells and squamous cell carcinoma.
Ritprajak P and Azuma M
Oral Oncology, 51(3), 221-228 (2015)
Integrative analysis reveals selective 9p24.1 amplification, increased PD-1 ligand expression, and further induction via JAK2 in nodular sclerosing Hodgkin lymphoma and primary mediastinal large B-cell lymphoma.
Green MR
Blood, 116(17), 3268-3277 (2010)
Adnan R Khan et al.
Nature communications, 6, 5997-5997 (2015-01-23)
Specific B-cell subsets can regulate T-cell immune responses, and are termed regulatory B cells (Breg). The majority of Breg cells described in mouse and man have been identified by IL-10 production and are known to suppress allergy and autoimmunity. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service