SRP6097
GSTA1 human
recombinant, expressed in E. coli, ≥90% (SDS-PAGE)
Synonym(s):
GST2, GSTA1-1, GTH1, Glutathione S-transferase alpha 1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
recombinant
expressed in E. coli
Assay
≥90% (SDS-PAGE)
form
liquid
mol wt
25.6 kDa
packaging
pkg of 100 μg
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... GSTA1(2938)
General description
Glutathione S-transferase A1 in humans is encoded by the GSTA1 gene. Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. These enzymes function in the detoxification of electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins and products of oxidative stress, by conjugation with glutathione. The genes encoding these enzymes are known to be highly polymorphic. GSTA1 protects the cells from reactive oxygen species and is highly expressed in liver.
The gene encoding GSTA1 is localized on human chromosome 6p12.2.
Biochem/physiol Actions
GSTA1 (Glutathione S-transferase α1) is associated with the biosynthesis of steroids such as testosterone and progesterone. It plays a vital role in double bond isomerase activity. GSTA1 converts δ(5)-androstene-3,17-dione to δ(4)-androstene-3,17-dione. It has a role in the metabolism of several xenobiotics.
Physical form
1 mg/mL solution in 20 mM Tris-HCl buffer (pH 8.0) containing 1 mM DTT and 10% glycerol.
Preparation Note
Centrifuge the vial prior to opening.
Other Notes
MAEKPKLHYF NARGRMESTR WLLAAAGVEF EEKFIKSAED LDKLRNDGYL MFQQVPMVEI DGMKLVQTRA ILNYIASKYN LYGKDIKERA LIDMYIEGIA DLGEMILLLP VCPPEEKDAK LALIKEKIKN RYFPAFEKVL KSHGQDYLVG NKLSRADIHL VELLYYVEEL DSSLISSFPL LKALKTRISN LPTVKKFLQP GSPRKPPMDE KSLEEARKIF RF
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Knockout and transgenic mice in glutathione transferase research.
Henderson CJ and Wolf CR
Drug Metabolism Reviews, 43(2), 152-164 (2011)
Crystal structure of human glutathione S-transferase A3-3 and mechanistic implications for its high steroid isomerase activity.
Yijun G, et al.
Biochemistry, 43(43) (2004)
Functional polymorphism of human glutathione transferase A3: effects on xenobiotic metabolism and steroid biosynthesis.
Tetlow N, et al.
Pharmacogenetics, 657-663 (2004)
Kaspars Tars et al.
Journal of molecular biology, 397(1), 332-340 (2010-01-20)
Glutathione transferases (GSTs) are abundant enzymes catalyzing the conjugation of hydrophobic toxic substrates with glutathione. In addition to detoxication, human GST A3-3 displays prominent steroid double-bond isomerase activity; e.g. transforming Delta(5)-androstene-3-17-dione into Delta(4)-androstene-3-17-dione (AD). This chemical transformation is a crucial
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service