Skip to Content
Merck
All Photos(1)

Key Documents

SRP4505

Sigma-Aldrich

FABP5 human

recombinant, expressed in E. coli

Synonym(s):

E-FABP, Epidermal-type fatty acid-binding protein, Fatty acid-binding protein 5, Fatty acid-binding protein epidermal, Psoriasis-associated fatty acid-binding protein homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE)

form

lyophilized

mol wt

~16 kDa

packaging

pkg of 100 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FABP5(2171)

General description

FABPs (fatty acid-binding proteins) are small 14-15kDa intracellular cytosolic proteins. They are needed for the transport of lipids to specific areas in cells.[1] The FABP5 gene is mapped to human chromosome 8q21.[2]

Biochem/physiol Actions

FABP5 (fatty acid-binding protein 5) is involved in the pathogenesis of metabolic diseases. Absence of FABP-4 and -5 in mice protects them against diet-induced obesity, insulin resistance, type 2 diabetes, atherosclerosis and fatty liver disease.[1] FABP5 is also associated with bladder, pancreas, prostate, breast cancers and glioblastoma.[3]

Physical form

Sterile filtered and Lyophilized without additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL. This solution can then be diluted into other aqueous buffers

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel fatty acid-binding protein (FABP) gene resulting from tandem gene duplication in mammals: transcription in rat retina and testis.
Liu RZ, et al.
Genomics, 92, 436-436 (2008)
Farzad S Forootan et al.
Oncotarget, 7(8), 9322-9339 (2016-01-28)
In previous work, it is suggested that the excessive amount of fatty acids transported by FABP5 may facilitate the malignant progression of prostate cancer cells through a FABP5-PPARγ-VEGF signal transduction axis to increase angiogenesis. To further functionally characterise the FABP5-PPARγ-VEGF
Fatty acid activated PPAR? promotes tumorigenicity of prostate cancer cells by up regulating VEGF via PPAR responsive elements of the promoter.
Forootan FS, et al.
Oncotarget, 7, 9322-9322 (2016)
A critical role of fatty acid binding protein 4 and 5 (FABP4/5) in the systemic response to fasting.
Syamsunarno MR, et al.
PLoS ONE, 8, e79386-e79386 (2013)
Long Pan et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 106, 68-76 (2018-06-30)
Tumor angiogenesis is an essential process for facilitating tumor growth and metastasis. Fatty acid binding protein 5(FABP5)is highly expressed in hepatocellular carcinoma (HCC). Thus, we investigated the role of FABP5 in tumor angiogenesis during HCC development. In this study, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service