Skip to Content
Merck
All Photos(1)

Key Documents

SML2631

Sigma-Aldrich

IITZ-01

≥98% (HPLC)

Synonym(s):

N2-[4-(1H-Bbenzimidazol-2-yl)phenyl]-N4-(4-fluorophenyl)-6-(4-morpholinyl)-1,3,5-triazine-2,4-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H23FN8O
CAS Number:
Molecular Weight:
482.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

IITZ-01 is a cell penetrant and potent autophagy inhibitor in numerous cancer cells including breast, lung, and colon cancer cells. It significantly elevates LC3-II and SQSTM1 protein levels in starvation-induced autophagy conditions. IITZ-01 accumulates within lysosomes causing liposomal deacidification, impairment of lysosomal maturation and inhibition of lysosomal degradation. Additionally, IITZ-01 induces apoptosis through the mitochondria-mediated pathway in TNBC cells MDA-MB-231 and MDA-MB-453, while sparing normal mammary epithelial MCF10A cell.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of new s-triazine bearing benzimidazole and benzothiazole derivatives as anticancer agents
Kumar GJ, Kumar SN, Thummuri D, Adari LBS, Naidu VGM, Srinivas K, Rao VJ
Medicinal Chemistry Research, 24, 3991-4001 (2015)
Lalita Guntuku et al.
Oncogene, 38(4), 581-595 (2018-09-01)
Autophagy is a homeostatic process that recycles damaged organelles and long-lived proteins by delivering them in double-membrane vesicles to lysosomes for degradation. Autophagy has a prominent role in survival, proliferation, and resistance of tumors in metabolic and chemotherapeutic stress conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service