Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200118

Sigma-Aldrich

Anti-CYBB antibody produced in rabbit

~1.5 mg/mL, affinity isolated antibody

Synonym(s):

Anti-CGD, Anti-Cytochrome b-245, β polypeptide (chronic granulomatous disease), Anti-GP91-1, Anti-GP91-PHOX, Anti-NOX2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~95 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1.5 mg/mL

technique(s)

western blot: 1.5-3.0 μg/mL using RAW264 cell extracts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYBB(1536)

Looking for similar products? Visit Product Comparison Guide

General description

CYBB (cytochrome b-245), belongs to the family of NADPH oxidases, that catalyze the generation of the superoxide ion. It is a six-transmembrane glycoprotein, that binds heme, flavin and NADPH. CYBB associates with membrane-bound p22phox to assemble the heavy subunit of flavocytochrome b558, the catalytic component of phagocyte NADPH oxidase. It also associates with four cytosolic components p47phox, p67phox, p40phox and Rac2 required for NADPH oxidase activity. Homologs of the NOX protein family include the catalytic subunits NOX1, NOX3-5, Duox1 and Duox2. NOX2/gp91phox is expressed in neutrophils and phagocytes.

Application

Anti-CYBB antibody produced in rabbit has been used in immunoblotting and western blotting.

Biochem/physiol Actions

CYBB (also known as cytochrome b-245) promotes neurotoxic activation of microglia, that play a central role during neuroinflammatory states and in amyotrophic lateral sclerosis (ALS). NOX1, NOX3-5, Duox1 and Duox2 plays important roles in redox-dependent cell signaling, inflammation and in neurodegenerative diseases. In ALS mice, deletion of either NOX1 or NOX2 gene has significantly slowed disease progression and improved survival.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shock Wave Therapy Enhances Mitochondrial Delivery into Target Cells and Protects against Acute Respiratory Distress Syndrome
Lin KC, et al.
Mediators of Inflammation, 2018 (2018)
Perinatal alpha-tocopherol overload programs alterations in kidney development and renal angiotensin II signaling pathways at birth and at juvenile age: Mechanisms underlying the development of elevated blood pressure
Ribeiro VS, et al.
Biochimica et Biophysica Acta (BBA)-Molecular Basis of Disease, 1864(7), 2458-2471 (2018)
Nox enzymes, ROS, and chronic disease: an example of antagonistic pleiotropy
Lambeth JD
Free Radical Biology & Medicine, 43(3), 332-347 (2007)
Kuan-Hung Chen et al.
Journal of neuropathology and experimental neurology, 78(1), 65-77 (2018-11-28)
This study tested the hypothesis that combined adipose-derived mesenchymal stem cell (ADMSC) and low-energy extracorporeal shock wave (ECSW) therapy could protect brain from brain death (BD)-induced injury. Adult male Sprague Dawley rats were categorized into group 1 (sham control), group
Ya Yue et al.
Stem cell research & therapy, 13(1), 186-186 (2022-05-08)
This experimental study was designed as a preclinical study for testing the hypothesis that intrarenal arterial (IRA) transfusion of human umbilical cord-derived mesenchymal stem cells (HUCDMSCs) therapy preserved the residual renal function of diabetic kidney disease (DKD) in rat [induction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service