Skip to Content
Merck
All Photos(1)

Key Documents

SAB1104866

Sigma-Aldrich

Anti-LY6H (54-62) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Lymphocyte antigen 6 complex, locus H

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen predicted mol wt ~15 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LY6H(4062)

General description

LY6H (lymphocyte antigen 6 family member H) belongs to the Ly6 family of genes and is mapped to the chromosome 8q24. The encoded protein is predominantly expressed in the brain and also expressed in pancreas, testis, small intestine and colon. Ly6h is a membrane bound protein and is localized to synaptic compartments.

Immunogen

synthetic peptide corresponding to amino acids 54-62 of human LY6H

Biochem/physiol Actions

LY6H (lymphocyte antigen 6 family member H) might be involved in central nervous system and the immune system.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation and Characterization of a New Member of the HumanLy6Gene Family (LY6H).
Horie M, et al.
Genomics, 53(3), 365-368 (1998)
Expression of the Ly-6 family proteins Lynx1 and Ly6H in the rat brain is compartmentalized, cell-type specific, and developmentally regulated.
Thomsen M S, et al.
Brain Structure &Amp; Function, 219(6), 1923-1934 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service