Recommended Products
form
solid
mol wt
~1300
color
white
SMILES string
OC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5COC(=O)CCC(O)=O)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]1[C@H](O)[C@H]2O
InChI
1S/C40H64O33/c41-3-9-29-17(49)23(55)35(62-9)69-30-10(4-42)64-37(25(57)19(30)51)71-32-12(6-44)66-39(27(59)21(32)53)73-34-14(8-61-16(48)2-1-15(46)47)67-40(28(60)22(34)54)72-33-13(7-45)65-38(26(58)20(33)52)70-31-11(5-43)63-36(68-29)24(56)18(31)50/h9-14,17-45,49-60H,1-8H2,(H,46,47)/t9-,10-,11-,12-,13-,14-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-/m1/s1
InChI key
MQYVFTUFHBQDRU-QHEXKRCRSA-N
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service