Skip to Content
Merck
All Photos(1)

Documents

S1013

Sigma-Aldrich

(−)-Scopolamine hydrochloride

≥90% (HPLC), powder

Synonym(s):

Hyoscine hydrochloride, Scopine tropate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO4 · HCl
CAS Number:
Molecular Weight:
339.81
Beilstein:
4168778
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥90% (HPLC)

form

powder

optical activity

[α]25/D ~−27°, c = 1 in H2O(lit.)

color

white

solubility

H2O: 50 mg/mL

SMILES string

Cl.[H][C@]12C[C@H](C[C@]([H])(N1C)[C@]3([H])O[C@]23[H])OC(=O)[C@H](CO)c4ccccc4

InChI

1S/C17H21NO4.ClH/c1-18-13-7-11(8-14(18)16-15(13)22-16)21-17(20)12(9-19)10-5-3-2-4-6-10;/h2-6,11-16,19H,7-9H2,1H3;1H/t11-,12-,13-,14+,15-,16+;/m1./s1

InChI key

KXPXJGYSEPEXMF-MOUKNHLCSA-N

Gene Information

human ... CHRM1(1128)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Competitive nonselective muscarinic acetylcholine antagonist. Scopolamine-induced amnesia in laboratory animals is a commonly-used model of memory deficit.

Caution

The powder is stable at least four years stored at room temperature.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stacey J Sukoff Rizzo et al.
Current protocols in mouse biology, 8(2), e45-e45 (2018-06-21)
The relationship between chronological age (lifespan) and biological age (healthspan) varies amongst individuals. Understanding the normal trajectory and characteristic traits of aging mice throughout their lifespan is important for selecting the most reliable and reproducible measures to test hypotheses. The
Sunjoo Joo et al.
Nature communications, 13(1), 1133-1133 (2022-03-05)
Most sexual organisms inherit organelles from one parent, commonly by excluding organelles from the smaller gametes. However, post-mating elimination of organelles derived from one gamete ensures uniparental inheritance, where the underlying mechanisms to distinguish organelles by their origin remain obscure.
X Bian et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 21(9), 969-977 (2009-04-21)
Neuromuscular transmission is crucial for normal gut motility but little is known about its postnatal maturation. This study investigated excitatory/inhibitory neuromuscular transmission in vitro using ileal nerve-muscle preparations made from neonatal (< or =48 h postnatal) and adult ( approximately
Fang-Fang Chen et al.
Oncoimmunology, 9(1), 1831153-1831153 (2020-10-29)
Therapeutic monoclonal antibodies against the PD-L1/PD-1 (programmed death ligand-1/programmed cell death protein-1) axis have achieved great successes in cancer treatments, but the development of small-molecule immunomodulators of the pathway has lagged far behind. We established a cellular coculture assay with
Narayana Krishna Yelleswarapu et al.
Neurogastroenterology and motility, 34(12), e14439-e14439 (2022-12-03)
Alterations in gastrointestinal (GI) function and the gut-brain axis are associated with progression and pathology of Alzheimer's Disease (AD). Studies in AD animal models show that changes in the gut microbiome and inflammatory markers can contribute to AD development in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service