Skip to Content
Merck
All Photos(1)

Key Documents

P9648

Sigma-Aldrich

2-Linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine

≥97% (GC), ≥99% (TLC)

Synonym(s):

PC, 1-Hexadecanoyl-2-(cis-9,12-octadecadienoyl)-sn-glycero-3-phosphocholine, 3-sn-Phosphatidylcholine, 2-linoleoyl-1-palmitoyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H80NO8P
CAS Number:
Molecular Weight:
758.06
Beilstein:
5209585
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
Pricing and availability is not currently available.

biological source

synthetic (organic)

Assay

≥97% (GC)
≥99% (TLC)

form

solid

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O

InChI

1S/C42H80NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h14,16,20-21,40H,6-13,15,17-19,22-39H2,1-5H3/b16-14-,21-20-/t40-/m1/s1

InChI key

JLPULHDHAOZNQI-ZTIMHPMXSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laurence Lins et al.
International journal of molecular sciences, 20(16) (2019-08-21)
Since the 50's, the massive and "environmental naïve" use of synthetic chemistry has revolutionized the farming community facing the dramatic growth of demography. However, nowadays, the controversy grows regarding the long-term harmful effects of these products on human health and
Aleksandra Stamenkovic et al.
American journal of physiology. Heart and circulatory physiology, 320(3), H1170-H1184 (2021-01-30)
Myocardial ischemia-reperfusion (I/R) injury increases the generation of oxidized phosphatidylcholines (OxPCs), which results in cell death. However, the mechanism by which OxPCs mediate cell death and cardiac dysfunction is largely unknown. The aim of this study was to determine the
Mehdi Ravandeh et al.
Biomolecules, 11(2) (2021-03-07)
In the eye lens cell membrane, the lipid composition changes during the aging process: the proportion of sphingomyelins (SM) increases, that of phosphatidylcholines decreases. To investigate the protective role of the SMs in the lens cell membrane against oxidative damage
Grigory G Borisenko et al.
Nucleic acids research, 37(4), e28-e28 (2009-01-23)
We report a novel approach for the attachment of DNA fragments to the surface of live cells. By using fluorescence microscopy and flow cytometry we demonstrated that our synthetic conjugates of fatty acid with oligonucleotides can be incorporated in plasma
Elise L Donovan et al.
Diabetology & metabolic syndrome, 5(1), 24-24 (2013-05-16)
Lipidomic analysis was performed to explore differences in lipid profiles between plasma from lean and obese subjects, followed by in vitro methods to examine a role for the identified lipids in endothelial cell pathophysiology. Plasma was collected from 15 morbidly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service