Skip to Content
Merck
All Photos(2)

Key Documents

P4621

Sigma-Aldrich

Anti-Potassium Channel Kir6.2 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-KCNJ11

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

western blot: 1:200 using rat pancreas membranes

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNJ11(3767)
mouse ... Kcnj11(16514)
rat ... Kcnj11(83535)

Specificity

This antibody is specific for Kir6.2 and does not cross-react with Kir6.1.

Immunogen

peptide corresponding to amino acid residues 372-385 of rat Kir6.2. This epitope is identical in mouse protein and highly homologous in human protein (12/14 residues identical).

Physical form

Lyophilized at ~0.6 mg/ml from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.025% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service