P3476
Protein A–Agarose Fast Flow
50%, aqueous suspension
Synonym(s):
Protein A resin
About This Item
Recommended Products
biological source
protein A from Staphylococcus aureus
description
protein A leaching <= 5 ng/mL
form
aqueous suspension
concentration
50%
matrix
6% Fast-flow beaded agarose
capacity
37-43 mg/mL binding capacity (IgG)
storage temp.
2-8°C
Application
Protein A–Agarose Fast Flow has been used:
- in the immunoprecipitation of protein complex TRRAP/p400 from viral transfected human normal kidney and baby rat kidney cells
- in the purification of polyclonal IgG from antisera
- in the immunoprecipitation assay of human embryonic kidney cells transfected by HA-Ano9 plasmids
Features and Benefits
- High capacity
- Low leakage
- Stable affinity medium for the purification of monoclonal and polyclonal antibodies
- Ideal for rapid capture of immunoglobulins from cell culture
Physical form
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Workflows for monoclonal antibody adalimumab characterization ensure drug safety and efficacy through critical quality attribute analysis.
Workflows for monoclonal antibody adalimumab characterization ensure drug safety and efficacy through critical quality attribute analysis.
Workflows for monoclonal antibody adalimumab characterization ensure drug safety and efficacy through critical quality attribute analysis.
Workflows for monoclonal antibody adalimumab characterization ensure drug safety and efficacy through critical quality attribute analysis.
Protocols
SEC-MS protocol for rapid glycoprofiling of monoclonal antibodies with antibody purification and mass spectrometer calibration.
SEC-MS protocol for rapid glycoprofiling of monoclonal antibodies with antibody purification and mass spectrometer calibration.
SEC-MS protocol for rapid glycoprofiling of monoclonal antibodies with antibody purification and mass spectrometer calibration.
SEC-MS protocol for rapid glycoprofiling of monoclonal antibodies with antibody purification and mass spectrometer calibration.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service