Skip to Content
Merck

M220

Monoclonal Anti-Nicotinic Acetylcholine Receptor (α7 Subunit) antibody produced in mouse

~5 mg/mL, clone mAb 306, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CHRNA7-2, Anti-NACHRA7

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
MDL number:
Conjugate:
unconjugated
Clone:
mAb 306, monoclonal
Application:
IHC (f)
Citations:
12
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

mAb 306, monoclonal

form

buffered aqueous solution

species reactivity

rat (denatured), human (denatured), chicken

concentration

~5 mg/mL

technique(s)

immunohistochemistry (frozen sections): 1:3,000

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHRNA7(1139)
rat ... Chrna7(25302)

General description

Nicotinic acetylcholine receptors (nAChRs) function as ligand-gated channels that modulate endoplasmic reticulum stress and neurotransmitter release. These receptors also control synaptic signaling in ganglia and muscles. The α7 form of nAChR is expressed in mitochondria and regulates Ca2+ transport and the release of cytochrome c. When expressed in non-excitable cells, α7 nAChR mediates cell growth and survival
Monoclonal anti-nicotinic acetylcholine receptor ((α7) subunit) antibody is specific for native and denatured α7 subunits in chick, as well as for the denatured α7 subunits in rats and humans. The antibody does not bind well to the native form of the mammalian α7 subunit. The product can be used in chick, rat and human tissues.

Immunogen

native and denatured α7 subunit (amino acids 380-400, binds α-bungarotoxin) of the nicotinic acetylcholine receptor from chicken and the denatured α7 subunit from rat.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
Monoclonal anti-nicotinic acetylcholine receptor ((α7) subunit) antibody is suitable for use in immunocytochemistry (at 1:3000-1:50 dilutions using paraformaldehyde fixed, cultured rat chromaffin cells) , and immunohistochemistry (at 1:3,000 dilutions using frozen sections).

Physical form

Solution in 20 mM Na3PO4, pH 7.2, containing 150 mM NaCl and 0.05% sodium azide.

Legal Information

Sold with permission of the Salk Institute

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library