KEM0001
Uracil DNA Glycosylase (UDG)
Ultra-pure enzyme for nucleic acid modifications
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
grade
Molecular Biology
for molecular biology
Assay
>99% (SDS-PAGE)
form
buffered aqueous solution
specific activity
77,000 U/mg
concentration
2,000 U/mL
shipped in
dry ice
storage temp.
−20°C
General description
Uracil-DNA Glycosylase catalyzes the hydrolysis of the N-glycosylic bond between the uracil and sugar, leaving an abasic site in uracil-containing single or double-stranded DNA. The enzyme shows no measurable activity on short oligonucleotides (<6 bases), or RNA substrates.
Application
Suitable for:
- Removing uracil from DNA
- Control of carry-over contamination in PCR
Features and Benefits
- Ultra-purification process for ultimate enzyme performance
- Highest quality specifications for ultimate product consistency
- Undetectable DNA and nuclease contamination
Components
Supplied with:KEM0001B (10X UDG Reaction Buffer)
Unit Definition
1 unit is defined as the amount of enzyme that catalyzes the release of 1.8 nmol of Uracil in 30 minutes from double-stranded, tritiated, Uracil containing-DNA at 37° C in 1X UDG Reaction Buffer.
Physical form
Supplied in 10 mM Tris-HCl, 50 mM NaCl, 1 mM DTT, 0.1 mM EDTA, and 50% glycerol at pH 7.5 @ 25° C
Other Notes
Source of protein: A recombinant E. coli strain carrying the Uracil DNA Glycosylase gene from E. coli K-12.
Unit Size: 10,000 U
related product
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Laure Rittié et al.
Journal of cell communication and signaling, 2(1-2), 25-45 (2008-09-04)
Since molecular cloning has become routine laboratory technique, manufacturers offer countless sources of enzymes to generate and manipulate nucleic acids. Thus, selecting the appropriate enzyme for a specific task may seem difficult to the novice. This review aims at providing
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service