Skip to Content
Merck
All Photos(1)

Documents

HPA002911

Sigma-Aldrich

Anti-AADAC antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Arylacetamide deacetylase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

PALQPLDVDLPSYQENSNFLFLSKSLMVRFWSEYFTTDRSLEKAMLSRQHVPVESSHLFKFINWSSLLPERFIKGHVYNNPNYGSSELAKKYPGFLDVRAAPLLADDNKLRGLPLTYVITCQYDLLRDDGLMYVTRLRNTGV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AADAC(13)

Immunogen

Arylacetamide deacetylase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Arylacetamide deacetylase is an enzyme encoded by the AADAC gene in humans. It is a glycoprotein and can hydrolyze clinical drugs, such as flutamide, phenacetin and rifamycins. The enzyme competes against the activity of cytosolic arylamine N-acetyltransferase, which catalyzes one of the initial biotransformation pathways for arylamine and heterocyclic amine carcinogens in many species and tissues. It is also responsible for the deacetylation of rifamycins and affects the induction rate of drug-metabolizing enzymes by rifamycins and their induced hepatotoxicity. It may be an important enzyme responsible for flutamide-induced hepatotoxicity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86503

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyotaka Muta et al.
Biochemical pharmacology, 87(2), 352-359 (2013-10-16)
Human arylacetamide deacetylase (AADAC) can hydrolyze clinical drugs such as flutamide, phenacetin, and rifamycins. AADAC is a glycoprotein, but the role of glycosylation remains unclear. In the present study, we investigated the effect of glycosylation on AADAC enzyme activity. Immunoblot
Mai Shimizu et al.
Drug metabolism and disposition: the biological fate of chemicals, 40(6), 1183-1190 (2012-03-15)
Human arylacetamide deacetylase (AADAC) is responsible for the hydrolysis of clinically used drugs such as flutamide, phenacetin, and rifamycins. Our recent studies suggested that human AADAC is a relevant enzyme pharmacologically and toxicologically. To date, the genetic polymorphisms that affect
Akinori Nakajima et al.
Biochemical pharmacology, 82(11), 1747-1756 (2011-08-23)
Rifamycins such as rifampicin, rifabutin, and rifapentine are used for the treatment of tuberculosis and induce various drug-metabolizing enzymes. Rifamycins have been reported to be mainly deacetylated by esterase(s) expressed in human liver microsomes (HLM) to 25-deacetylrifamycins, but the responsible
Akinobu Watanabe et al.
Drug metabolism and disposition: the biological fate of chemicals, 37(7), 1513-1520 (2009-04-03)
Flutamide, an antiandrogen drug, is widely used for the treatment of prostate cancer. The initial metabolic pathways of flutamide are hydroxylation and hydrolysis. It was recently reported that the hydrolyzed product, 4-nitro-3-(trifluoromethyl)phenylamine (FLU-1), is further metabolized to N-hydroxy FLU-1, an
M R Probst et al.
The Journal of biological chemistry, 269(34), 21650-21656 (1994-08-26)
Microsomal arylacetamide deacetylase (DAC) competes against the activity of cytosolic arylamine N-acetyltransferase, which catalyzes one of the initial biotransformation pathways for arylamine and heterocyclic amine carcinogens in many species and tissues. Activity determination and immunoblot analysis of DAC in human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service