Skip to Content
Merck
All Photos(2)

Key Documents

H6258

Sigma-Aldrich

Hydroxyalkoxypropyl-Dextran

Type VI

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817

type

Type VI

Quality Level

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Hydroxyalkoxypropyl-Dextran is used for liquid chromatography, protein chromatography, and gel filtration chromatography. Hydroxyalkoxypropyl-Dextran has been used to develop a nondestructive method for effective removal of lipids from small quantities of aqueous protein as well as to develop a highly specific method for the determination of cortisol in human serum and urine.
Lipophilic, hydrophobic gel for liquid chromatography.

Other Notes

Hydroxypropyl beaded dextran (Lipophilic Sephadex, LH-20-100), substituted with long chain alkyl ethers.
Substituted approx. 10% by weight with alkyl chains of length C15-C18

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gregory P Tochtrop et al.
Biochemistry, 42(40), 11561-11567 (2003-10-08)
Human ileal bile acid binding protein (I-BABP) is a member of the intracellular lipid binding protein family. This protein is thought to function in the transcellular transport and enterohepatic circulation of bile salts. Human I-BABP binds two molecules of glycocholate
Gergő Horváth et al.
The FEBS journal, 283(3), 541-555 (2015-11-28)
Besides aiding digestion, bile salts are important signal molecules exhibiting a regulatory role in metabolic processes. Human ileal bile acid binding protein (I-BABP) is an intracellular carrier of bile salts in the epithelial cells of the distal small intestine and
Gregory P Tochtrop et al.
Journal of the American Chemical Society, 126(35), 11024-11029 (2004-09-02)
The recognition between proteins and their native ligands is fundamental to biological function. In vivo, human ileal bile acid binding protein (I-BABP) encounters a range of bile salts that vary in the number and position of steroidal hydroxyl groups and
Mariana Suárez et al.
PloS one, 15(11), e0242312-e0242312 (2020-11-13)
The intestinal fatty acid binding protein (FABP) is a small protein expressed along the small intestine that bind long-chain fatty acids and other hydrophobic ligands. Several lines of evidence suggest that, once in the nucleus, it interacts with nuclear receptors
Christophe Anjard et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(21), 7607-7611 (2005-05-18)
A wide variety of mechanisms have evolved for intercellular communication in metazoans, but some of the signaling molecules were already used in their predecessors. The social amoeba, Dictyostelium discoideum, is known to use peptides to trigger sporulation within fruiting bodies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service