All Photos(1)
About This Item
Linear Formula:
HOC6H4CH2CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein:
1448766
EC Number:
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22
Recommended Products
mp
148-151 °C (lit.)
SMILES string
OC(=O)Cc1ccc(O)cc1
InChI
1S/C8H8O3/c9-7-3-1-6(2-4-7)5-8(10)11/h1-4,9H,5H2,(H,10,11)
InChI key
XQXPVVBIMDBYFF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service