Skip to Content
Merck
All Photos(6)

Key Documents

CWA-1121

Sigma-Aldrich

Anti-Mouse CD11a (M17/4) ColorWheel® Dye-Ready mAb

greener alternative

for use with ColorWheel® Dyes (Required, (Sold Separately))

Synonym(s):

Integrin alpha-L;CD11 antigen-like family member A;Leukocyte adhesion glycoprotein LFA-1 alpha chain;LFA-1A;Leukocyte function-associated molecule 1 alpha chain;Lymphocyte antigen 15;Ly-15

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

M17/4, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 128.33 kDa

purified by

using Protein A

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable

isotype

IgG2aκ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

compatibility

for use with ColorWheel® Dyes (Required, (Sold Separately))

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Itgal(16408)

General description

ColorWheelColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheelColorWheel® antibody with any ColorWheelColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheelColorWheel® antibody and dye is lyophilized for long-term storage, allowing for simplicity and flexibility without compromise. ColorWheelColorWheel® technology requires both ColorWheelColorWheel® antibody and ColorWheelColorWheel® dye for Flow Cytometry application.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Specificity

Clone M17/4 is a rat monoclonal antibody that detects murine CD11a (Integrin aL). It targets an epitope within the extracellular domain.

Immunogen

C57BL/6 Mouse Splenic Secondary Cytotoxic T Lymphocytes.

Application

Quality Control Testing

Evaluated by Flow Cytometry in C57BL/6 mouse splenocytes.

Flow Cytometry Analysis:

Staining of one million C57BL/6 mouse splenocytes was performed using 5 μL of a 1:1 mixture of Cat. No. CWA-1121, Anti-Mouse CD11a (M17/4) ColorWheelColorWheel® Dye-Ready mAb and Cat. No. CWDS-PE ColorWheelColorWheel® Antibody-Ready Phycoerythrin (PE) Dye or an equivalent amount of Cat. No. CWA-1018 Rat IgG2a isotype control and Cat. No. CWDS-PE ColorWheelColorWheel® Antibody-Ready Phycoerythrin (PE) Dye.

Tested Applications

Flow Cytometry Analysis: Staining of one million C57BL/6 mouse splenocytes was performed using 5 μL of a 1:1 mixture of Cat. No. CWA-1121, Anti-Mouse CD11a (M17/4) ColorWheelColorWheel® Dye-Ready mAb and Cat. No. CWDS-APC ColorWheelColorWheel® Antibody-Ready Allophycocyanin (APC) Dye or an equivalent amount of Cat. No. CWA-1018 Rat IgG2a isotype control and Cat. No. CWDS-APC ColorWheelColorWheel® Antibody-Ready Allophycocyanin (APC) Dye.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Integrin alpha-L (UniProt: P24063; also known as CD11 antigen-like family member A, Leukocyte adhesion glycoprotein LFA-1 alpha chain, LFA-1A, Leukocyte function-associated molecule 1 alpha chain, Lymphocyte antigen 15, Ly-15, CD11a) is encoded by the Itgal (also known as Lfa-1, Ly-15) gene (Gene ID: 16408) in murine species. Integrins are part of a family of heterodimeric transmembrane glycoproteins that recognize a variety of ligands and control numerous physiologic functions in cells. CD11a is a single-pass type I membrane glycoprotein protein that is synthesized with a signal peptide (aa 1-23), which is subsequently cleaved off in the mature form that contains a large extracellular domain (aa 24-1087), a transmembrane domain (aa 1088-1108), and a cytoplasmic domain 1109-1163). CD11a noncovalently associates with b2 integrin to forms a heterodimer to serve as a receptor for ICAM1, ICAM2, ICAM3 and ICAM4. It is expressed on B and T lymphocytes, monocytes, macrophages, neutrophils, basophils, and eosinophils. It is also involved in generation of common lymphoid progenitor cells in bone marrow. In resting T-cells, about 40% of CD11a is constitutively phosphorylated and phosphorylation causes conformational changes needed for ligand binding and is considered to be essential for activation by some physiological agents. CD11a -/- mice exhibit decreased thymic cellularity and impaired early T cell development. CD11a is shown to be critical for generation of common lymphoid progenitors (CLPs) and lymphoid-primed multipotent progenitors (LMPPs). (Ref.: Bose, TO., et al. (2014). J. Immunol. 193(6); 2863-2872).

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Reconstitution

1.0 mg/mL after reconstitution at 25 μL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 μL PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

required but not provided

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service