APREST95174
PrEST Antigen POU2F3
Prestige Antibodies® Powered by Atlas Antibodies, buffered aqueous solution
Synonym(s):
Epoc-1, OCT11, PLA-1, Skn-1a
About This Item
Recommended Products
recombinant
expressed in E. coli
product line
Prestige Antibodies® Powered by Atlas Antibodies
Assay
>80% (SDS-PAGE)
form
buffered aqueous solution
mol wt
predicted mol wt 24 kDa
purified by
immobilized metal affinity chromatography (IMAC)
concentration
≥0.5 mg/mL
sequence note
PVHSTMPGTVTSSCSPGNNSRPSSPGSGLHASSPTASQNNSKAAVNSASSFNSSGSWYRWNHSTYL
Ensembl | human accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... POU2F3(25833)
General description
Application
Linkage
Physical form
Storage and Stability
Other Notes
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service