Skip to Content
Merck

Skip To

A3759

p-Aminohippuric acid sodium salt

Synonym(s):

N-(p-Aminobenzoyl)glycine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C9H9N2O3Na
CAS Number:
Molecular Weight:
216.17
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352106
EC Number:
202-309-8
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (TLC)

form

powder

color

white to off-white

application(s)

cell analysis

SMILES string

[Na].Nc1ccc(cc1)C(=O)NCC(O)=O

InChI

1S/C9H10N2O3.Na.H/c10-7-3-1-6(2-4-7)9(14)11-5-8(12)13;;/h1-4H,5,10H2,(H,11,14)(H,12,13);;

InChI key

IGACXVXMVJUFOL-UHFFFAOYSA-N

Biochem/physiol Actions

Para-Aminohippuric acid (PAH), OAT-1 specific, is used to help differentiate and characterized organic anion transporter (OAT) isoforms.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters