Skip to Content
Merck
All Photos(1)

Key Documents

75048

Sigma-Aldrich

Hyaluronic acid sodium salt from Streptococcus equi

mol wt 70,000-90,000

Synonym(s):

Sodium hyaluronate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

form

powder

mol wt

70,000-90,000

suitability

conforms to structure for Infrared spectrum

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C28H44N2O23.Na/c1-5(33)29-9-18(11(35)7(3-31)47-25(9)46)49-28-17(41)15(39)20(22(53-28)24(44)45)51-26-10(30-6(2)34)19(12(36)8(4-32)48-26)50-27-16(40)13(37)14(38)21(52-27)23(42)43;/h7-22,25-28,31-32,35-41,46H,3-4H2,1-2H3,(H,29,33)(H,30,34)(H,42,43)(H,44,45);/q;+1/t7-,8-,9-,10-,11-,12-,13+,14+,15-,16-,17-,18-,19-,20+,21+,22+,25-,26+,27-,28-;/m1./s1

InChI key

YWIVKILSMZOHHF-QJZPQSOGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

suitable as substrate for hyaluronidase

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Florian Rehfeldt et al.
Integrative biology : quantitative biosciences from nano to macro, 4(4), 422-430 (2012-02-22)
Physical features of microenvironments such as matrix elasticity E can clearly influence cell morphology and cell phenotype, but many differences between model matrices raise questions as to whether a standard biological scale for E exists, especially in 3D as well
Stephanie K Seidlits et al.
Biomaterials, 31(14), 3930-3940 (2010-02-23)
We report the ability to direct the differentiation pathway of neural progenitor cells (NPCs) within hydrogels having tunable mechanical properties. By modifying the polymeric sugar hyaluronic acid (HA), a major extracellular matrix component in the fetal mammalian brain, with varying
Mario W Kramer et al.
Cancer, 117(6), 1197-1209 (2010-10-21)
Cancer biomarkers are the backbone for the implementation of individualized approaches to bladder cancer (BCa). Hyaluronic acid (HA) and all 7 members of the HA family, that is, HA synthases (HA1, HA2, HA3), HYAL-1 hyaluronidase, and HA receptors (CD44s, CD44v
Ki Young Choi et al.
Biomaterials, 31(1), 106-114 (2009-09-29)
Hyaluronic acid nanoparticles (HA-NPs), which are formed by the self-assembly of hydrophobically modified HA derivatives, were prepared to investigate their physicochemical characteristics and fates in tumor-bearing mice after systemic administration. The particle sizes of HA-NPs were controlled in the range
Jason A Burdick et al.
Advanced materials (Deerfield Beach, Fla.), 23(12), H41-H56 (2011-03-12)
Hyaluronic acid (HA), an immunoneutral polysaccharide that is ubiquitous in the human body, is crucial for many cellular and tissue functions and has been in clinical use for over thirty years. When chemically modified, HA can be transformed into many

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service