Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C23H34N2O4S
CAS Number:
Molecular Weight:
434.59
PubChem Substance ID:
UNSPSC Code:
12352200
Beilstein/REAXYS Number:
8946533
MDL number:
Pricing and availability is not currently available.
product line
BioReagent
assay
≥98.0% (TLC)
solubility
DMSO: soluble, methanol: soluble
fluorescence
λex 335 nm; λem 516 nm in methanol
suitability
suitable for fluorescence
storage temp.
2-8°C
SMILES string
CN(C)c1cccc2c(cccc12)S(=O)(=O)NCCCCCCCCCCC(O)=O
InChI
1S/C23H34N2O4S/c1-25(2)21-15-11-14-20-19(21)13-12-16-22(20)30(28,29)24-18-10-8-6-4-3-5-7-9-17-23(26)27/h11-16,24H,3-10,17-18H2,1-2H3,(H,26,27)
InChI key
CEPGVMDMVJGHFQ-UHFFFAOYSA-N
Application
11-[5-(Dimethylamino)-1-naphthalenesulfonylamino]undecanoic acid (DAUDA), a dansyl-type fluorophore, is widely used as a hydrophobic binding site probe to identify and characterize the binding sites of medium chain fatty acids, their derivatives and other hydrophobic compounds on molecules such as proteins.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
PPE (personal protective equipment)
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
