Skip to Content
Merck
All Photos(1)

Key Documents

P4633

Sigma-Aldrich

Phenol Red

pH indicator

Synonym(s):

Phenolsulfonphthalein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14O5S
CAS Number:
Molecular Weight:
354.38
Beilstein:
326470
EC Number:
MDL number:
UNSPSC Code:
12171500

Pricing and availability is not currently available.

technique(s)

titration: suitable

visual transition interval

6.8-8.2, yellow to red

SMILES string

Oc1ccc(cc1)C2(OS(=O)(=O)c3ccccc23)c4ccc(O)cc4

InChI

1S/C19H14O5S/c20-15-9-5-13(6-10-15)19(14-7-11-16(21)12-8-14)17-3-1-2-4-18(17)25(22,23)24-19/h1-12,20-21H

InChI key

BELBBZDIHDAJOR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for use as a pH indicator.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hirotaka Miyamoto et al.
The Journal of pharmacy and pharmacology, 64(6), 848-854 (2012-05-11)
The aim of this study was to determine the factor changing the hepatic disposition of a drug during hypothermia using a rat liver perfusion system. The livers of male Wistar rats were perfused at 37, 32 or 28°C in the
Chromoendoscopy for H. pylori infection--time for a reaction?
José Carlos Marín-Gabriel et al.
Revista espanola de enfermedades digestivas : organo oficial de la Sociedad Espanola de Patologia Digestiva, 104(1), 1-3 (2012-02-04)
Dongdong Wang et al.
Journal of ethnopharmacology, 139(1), 189-193 (2011-11-22)
Bulbus Fritillaria Cirrhosae (BFC), known by the Chinese name "Chuan-Bei-Mu", is used as an antitussive, antiasthmatic and expectorant drug for more than 2000 years in China, and Bulbus of Fritillaria wabuensis S. Y. Tang & S. C. Yueh (BFW) was
Maki Nomura et al.
American journal of physiology. Renal physiology, 302(12), F1640-F1649 (2012-03-16)
Organic anion transporters (OAT1 and OAT3) and multidrug resistance-associated proteins (MRP2 and MRP4) play important roles in anionic drug secretion in renal proximal tubules. Changes in the expression of such transporters are considered to affect the tubular secretion of anionic
Jian-xiang Liu et al.
Zhejiang da xue xue bao. Yi xue ban = Journal of Zhejiang University. Medical sciences, 41(4), 430-433 (2012-08-29)
To develop a method for simultaneous assay of propulsion and absorption in small intestine. The mice were administrated through gastric tube with mixed reagents containing 0.12% phenol red, D-xylose (1.25%, 2.5% and 5%) and 15% gelatin. The influence of phenol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service