Skip to Content
Merck
All Photos(1)

Key Documents

92600

Supelco

Nobiletin

analytical standard

Synonym(s):

2-(3,4-Dimethoxyphenyl)-5,6,7,8-tetramethoxy-4H-1-benzopyran-4-one, 3′,4′,5,6,7,8-Hexamethoxyflavone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22O8
CAS Number:
Molecular Weight:
402.39
Beilstein:
360887
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water

application(s)

food and beverages

format

neat

SMILES string

COc1ccc(cc1OC)C2=CC(=O)c3c(OC)c(OC)c(OC)c(OC)c3O2

InChI

1S/C21H22O8/c1-23-13-8-7-11(9-15(13)24-2)14-10-12(22)16-17(25-3)19(26-4)21(28-6)20(27-5)18(16)29-14/h7-10H,1-6H3

InChI key

MRIAQLRQZPPODS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Nobiletin suppresses the expression of matrix metalloproteinases 1, 3 and 9, which participate in the break down of the extracellular matrix during tumor metastasis. Nobiletin is also involved in the increased expression of tissue inhibitor of matrix metalloproteinase 1 further preventing extracellular matrix degradation during tumor invasion. This bioflavonoid is believed to achieve both of these functions by disrupting the signal transduction pathway of Ras-Raf-MEK.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoru Horigome et al.
Bioscience, biotechnology, and biochemistry, 78(5), 851-860 (2014-07-19)
The rhizome of Kaempferia parviflora has been used in traditional Thai medicine. In this study, we identified and compared specific compounds from the hexane extract of K. parviflora with those from other Zingiberaceous plants by using gas chromatography-mass spectrometry. We
Feng-Quan Tan et al.
BMC plant biology, 15, 89-89 (2015-04-08)
Polyploidy has often been considered to confer plants a better adaptation to environmental stresses. Tetraploid citrus rootstocks are expected to have stronger stress tolerance than diploid. Plenty of doubled diploid citrus plants were exploited from diploid species for citrus rootstock
Jianchu Chen et al.
BMC pharmacology & toxicology, 15, 59-59 (2014-10-25)
Nobiletin is a non-toxic dietary flavonoid that possesses anti-cancer properties. Nobiletin has been reported to reduce the risk of prostate cancer, but the mechanism is not well understood. In this study, we investigated the effects of nobiletin in prostate cancer
Su-Chen Ho et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 71, 176-182 (2014-06-24)
Inhibiting microglial activation-mediated neuroinflammation has become a convincing target for the development of functional foods to treat neurodegenerative diseases. Tangerine peel (Citri reticulatae pericarpium) has potent anti-inflammatory capacity; however, its anti-neuroinflammatory capacity and the corresponding active compounds remain unclear. To
Adriana Arigò et al.
Journal of chromatography. A, 1649, 462183-462183 (2021-05-27)
In the European Union, cosmetic products are regulated by the Cosmetics Regulation (EC) No. 1223/2009. The Analytical Working Group of the International Fragrance Association (IFRA) suggested that a new sensitive analytical method is needed to determine psoralens in finished cosmetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service