Skip to Content
Merck
All Photos(1)

Documents

87824

Sigma-Aldrich

7-(Diethylamino)coumarin-3-carboxylic imidazolide

purum, fluorescent marker, ≥97.0% (N)

Synonym(s):

1-[7-(Diethylamino)-3-coumarinylcarbonyl]imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17N3O3
CAS Number:
Molecular Weight:
311.34
Beilstein:
8632400
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

purum

Assay

≥97.0% (N)

fluorescence

λex 385 nm; λem 574 nm in 0.1 M Tris pH 9.0

SMILES string

CCN(CC)c1ccc2C=C(C(=O)Oc2c1)C(=O)n3ccnc3

Other Notes

Reagent for the mild fluorescent labeling of hydroxy compounds

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lo et al.
Organic letters, 2(5), 683-685 (2000-05-18)
[reaction: see text] Imidazolide 1 has been prepared from 7-diethylaminocoumarin-3-carboxylic acid (2) and 1,1'-carbonyldiimidazole. This novel fluorescent derivatization reagent can be used in CD exciton chirality method for microscale structural determinations of hydroxyl-containing compounds. It features a red-shifted chromophore and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service