Skip to Content
Merck
All Photos(1)

Key Documents

73788

Supelco

1-Nitropropane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2NO2
CAS Number:
Molecular Weight:
89.09
Beilstein:
506236
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

3.1 (vs air)

vapor pressure

7.5 mmHg ( 20 °C)

Assay

≥99.7% (GC)

autoignition temp.

788 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.401 (lit.)
n20/D 1.401

bp

131-132 °C (lit.)

density

0.998 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CCC[N+]([O-])=O

InChI

1S/C3H7NO2/c1-2-3-4(5)6/h2-3H2,1H3

InChI key

JSZOAYXJRCEYSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M L Cunningham et al.
Toxicology and applied pharmacology, 110(3), 505-513 (1991-09-15)
2-Nitropropane (2-NP) is mutagenic in a number of short-term mutagenicity assays in vitro and in vivo, and is a potent hepatocarcinogen in rats. A structural isomer, 1-nitropropane (1-NP), is mutagenic in V79 cells and can induce unscheduled DNA synthesis in
J Ong et al.
European journal of pharmacology, 264(1), 49-54 (1994-10-13)
In the guinea-pig isolated ileum, both baclofen (5-100 microM) and gamma-aminobutyric acid (GABA; 5-100 microM) induced a bicuculline-insensitive depression of cholinergic twitch contractions which was reversibly and competitively antagonised by 3-amino-1-nitropropane (50-250 microM). 3-Amino-1-nitropropane (pA2 = 5.0 +/- 0.1) was
I Linhart et al.
Chemico-biological interactions, 80(2), 187-201 (1991-01-01)
Unlike primary nitroalkanes, such as 1-nitropropane, the secondary nitroalkane 2-nitropropane is geno- and hepatotoxic. Nitroalkanes exist in equilibrium with alkane nitronates. In order to investigate the relationship between nitroalkane toxicity and generation and stability of nitronates, propane 1- or 2-nitronate
E George et al.
Carcinogenesis, 10(12), 2329-2334 (1989-12-01)
2-Nitropropane (2-NP) is a rat liver carcinogen, whilst the 1-isomer is non-carcinogenic in rodents. Although DNA repair tests in the rat liver discriminated clearly between the carcinogenic and the non-carcinogenic isomer, uniformly negative results have been published for the mouse
E S Fiala et al.
Carcinogenesis, 8(12), 1947-1949 (1987-12-01)
1-Nitropropane (1-NP), 2-nitropropane (2-NP), 1-azoxypropane (1-AP) and 2-azoxypropane (2-AP), were assayed for carcinogenicity by gavage in male Sprague-Dawley rats. 2-NP was given at 1 mmol/kg three times per week for 16 weeks. 1-NP (1 mmol/kg), 1-AP (0.1 mmol/kg), 2-AP (0.1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service