Skip to Content
Merck
All Photos(1)

Key Documents

73529

Supelco

Clobetasol

pharmaceutical impurity standard, ≥95.0% (HPLC)

Synonym(s):

Clobetasol propionate impurity G (PhEur), (11β,16β)-21-Chloro-9-fluoro-11,17-dihydroxy-16-methylpregna-1,4-diene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28ClFO4
CAS Number:
Molecular Weight:
410.91
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% related substances
≤5.0% solvents
≤5.0% water

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CCl

InChI

1S/C22H28ClFO4/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,24)17(26)10-20(16,3)22(12,28)18(27)11-23/h6-7,9,12,15-17,26,28H,4-5,8,10-11H2,1-3H3/t12-,15-,16-,17-,19-,20-,21-,22-/m0/s1

InChI key

FCSHDIVRCWTZOX-DVTGEIKXSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Repr. 1B - STOT RE 2

Target Organs

Adrenal gland,Immune system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Yawalkar et al.
Journal of the American Academy of Dermatology, 25(6 Pt 2), 1137-1144 (1991-12-01)
Both halobetasol propionate and clobetasol 17-propionate exerted very marked antiinflammatory, antiproliferative, and vasoconstrictive effects during evaluation in a range of dermatopharmacologic models. Halobetasol propionate was distinctly more potent than clobetasol 17-propionate in the ultraviolet-induced dermatitis inhibition assay in guinea pigs
Kai-Li Mao et al.
Colloids and surfaces. B, Biointerfaces, 160, 704-714 (2017-10-17)
A poor percutaneous penetration capability for most topical anti-inflammatory drugs is one of the main causes compromising their therapeutic effects on psoriatic skin. Even though curcumin has shown a remarkable efficacy in the treatment of psoriasis, its effective penetration through
Reese Imhof et al.
Clinical, cosmetic and investigational dermatology, 13, 897-910 (2020-12-10)
Frontal fibrosing alopecia (FFA) is a primary lymphocytic cicatricial alopecia that is often considered a clinical variant of lichen planopilaris (LPP) due to their shared histopathologic features. FFA is characterized by the recession of the frontal, temporal, or frontotemporal hairline;
E A Olsen et al.
Journal of the American Academy of Dermatology, 15(2 Pt 1), 246-255 (1986-08-01)
Clobetasol-17-propionate, the most potent of currently available topical steroids as predicted by the vasoconstrictor assay, has just been approved in the United States. In psoriasis, it has proved significantly more effective than class II steroids and as or more effective
Jason E Ham et al.
Cutaneous and ocular toxicology, 38(2), 112-117 (2018-05-04)
Formaldehyde allergic contact dermatitis (ACD) may be due to products with free formaldehyde or formaldehyde-releasing agents; however, assessment of formaldehyde levels in such products is infrequently conducted. The present study quantifies total releasable formaldehyde from "in-use" products associated with formaldehyde

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service