Skip to Content
Merck
All Photos(1)

Key Documents

63969

Supelco

2-Nonanone

analytical standard

Synonym(s):

Heptyl methyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6COCH3
CAS Number:
Molecular Weight:
142.24
Beilstein:
1743645
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
39021224
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.420
n20/D 1.421 (lit.)

bp

192 °C/743 mmHg (lit.)

mp

−21 °C (lit.)

density

0.82 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCCC(C)=O

InChI

1S/C9H18O/c1-3-4-5-6-7-8-9(2)10/h3-8H2,1-2H3

InChI key

VKCYHJWLYTUGCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

T D Dreesen et al.
The Biochemical journal, 203(1), 69-75 (1982-04-01)
Individual turbinals from the right and left sides of dog olfactory tissue were removed and nerve-ending-particle preparations were prepared. (Na+ + K+)-dependent ATPase activities of the individual preparations, and the effect of several odorous compounds [including (+)- and (-)-carvone] on
E R Troemel et al.
Cell, 91(2), 161-169 (1997-11-05)
Different olfactory cues elicit distinct behaviors such as attraction, avoidance, feeding, or mating. In the nematode C. elegans, these cues are sensed by a small number of olfactory neurons, each of which expresses several different odorant receptors. The type of
J A Dunn et al.
Experimental & applied acarology, 37(1-2), 131-140 (2005-09-24)
Two types of trap, the BT Trap and PC(TM) pitfall cone trap, were evaluated against the most widely used method of sampling and sieving for their ability to detect low numbers of storage mites (Acarus siro, Tyrophagus longior and Lepidoglyphus
D A Birnby et al.
Genetics, 155(1), 85-104 (2000-05-03)
Caenorhabditis elegans daf-11 and daf-21 mutants share defects in specific chemosensory responses mediated by several classes of sensory neurons, indicating that these two genes have closely related functions in an assortment of chemosensory pathways. We report that daf-11 encodes one
Janina Kühn et al.
Journal of agricultural and food chemistry, 55(9), 3599-3604 (2007-04-07)
Interactions of the model flavor compound 2-nonanone with individual milk proteins, whey protein isolate (WPI), and sodium caseinate in aqueous solutions were investigated. A method to quantify the free 2-nonanone was developed using headspace solid-phase microextraction followed by gas chromatography

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service