Skip to Content
Merck
All Photos(1)

Key Documents

62933

Supelco

Rubusoside

analytical standard

Synonym(s):

(4α)-13-(β-D-glucopyranosyloxy)kaur-16-en-18-oic acid β-D-glucopyranosyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H50O13
CAS Number:
Molecular Weight:
642.73
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% water

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

C[C@@]12CCC[C@](C)([C@H]1CC[C@]34CC(=C)[C@](CC[C@@H]23)(C4)O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)C(=O)O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O

InChI

1S/C32H50O13/c1-15-11-31-9-5-18-29(2,7-4-8-30(18,3)28(41)44-26-24(39)22(37)20(35)16(12-33)42-26)19(31)6-10-32(15,14-31)45-27-25(40)23(38)21(36)17(13-34)43-27/h16-27,33-40H,1,4-14H2,2-3H3/t16-,17-,18+,19+,20-,21-,22+,23+,24-,25-,26+,27+,29-,30-,31-,32+/m1/s1

InChI key

YWPVROCHNBYFTP-OSHKXICASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Componentof stevia.
Rubusoside is a transfructosylated steviol glycoside, which can be isolated particularly from the leaves of Rubus suavissimus.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoki Sugimoto et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 43(4), 250-253 (2002-11-20)
The main and minor constituents of tenryocha extract product, a natural sweetener, were investigated as a part of an ongoing study to evaluate its quality and safety as a food additive. Four constituents, namely rubusoside, steviolmonoside, panicloside IV, and ent-16
Fang Zhang et al.
Journal of pharmaceutical sciences, 100(7), 2778-2789 (2011-02-12)
Curcumin (CUR) is an active food compound, but its insolubility and instability in water contributes to low bioavailability. In this study, the solubility of CUR was enhanced by utilizing the solubilizing properties of rubusoside (RUB). The solubility of CUR in
Guixin Chou et al.
Journal of agricultural and food chemistry, 57(3), 1076-1083 (2009-01-14)
The quality of botanical food is increasingly assessed by the content of multiple bioactive compounds. This study reports, for the first time, an HPLC fingerprinting method for the quality evaluation of Rubus suavissimus leaves possessing multiple bioactivities. Five constituents, gallic
C M Compadre et al.
Biomedical & environmental mass spectrometry, 15(4), 211-222 (1988-02-15)
Steviol (ent-13-hydroxykaur-16-en-19-oic acid), the aglycone of various plant-derived glycoside sweeteners consumed by human populations, is known to be mutagenic toward Salmonella tymphimurium strain TM677 when metabolically activated using a 9000 x g supernatant fraction derived from the liver of Aroclor
K Ohtani et al.
Chemical & pharmaceutical bulletin, 39(12), 3172-3174 (1991-12-01)
1,4-alpha-Glucosylation at the 13-O-glycosyl moiety of stevioside (S) and rubusoside (RU) results in a significant increase of sweetness. Saponification of the 19-COO-beta-glucosyl linkage of S and RU yielded steviolbioside (SB) (= 13-O-beta-sophorosyl-steviol) and steviolmonoside (SM) (= 13-O-beta-glucosyl-steviol), respectively, both of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service