61419
Isopimpinellin
analytical standard
Synonym(s):
4,9-Dimethoxy-7H-furo[3,2-g][1]benzopyran-7-one, 4,9-Dimethoxyfuro[3,2-g]chromen-7-one, 5,8-Dimethoxy-6,7-furanocoumarin, 5,8-Dimethoxypsoralen, Isoimpinellin, NSC 217988, NSC 401288
About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥95.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
impurities
≤5.0% water
application(s)
food and beverages
format
neat
storage temp.
2-8°C
SMILES string
O=C1OC2=C(OC)C3=C(C=CO3)C(OC)=C2C=C1
InChI
1S/C13H10O5/c1-15-10-7-3-4-9(14)18-12(7)13(16-2)11-8(10)5-6-17-11/h3-6H,1-2H3
InChI key
DFMAXQKDIGCMTL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Recommended products
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service