The purity of this product was determined by Gas Chromatographic analysis (GC). This product has been discontinued.
55321
Hexamethylenetetramine
Synonym(s):
E239, 1,3,5,7-Tetraazatricyclo[3.3.1.13,7]decane, Hexamine, Methenamine, Urotropine
About This Item
Recommended Products
grade
analytical standard
Quality Level
vapor pressure
<0.01 mmHg ( 20 °C)
Assay
≥99.0% (GC)
autoignition temp.
770 °F
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
impurities
≤1.0% water
mp
280 °C (subl.) (lit.)
solubility
H2O: soluble
application(s)
cleaning products
cosmetics
food and beverages
personal care
format
neat
SMILES string
C1N2CN3CN1CN(C2)C3
InChI
1S/C6H12N4/c1-7-2-9-4-8(1)5-10(3-7)6-9/h1-6H2
InChI key
VKYKSIONXSXAKP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 2 - Skin Sens. 1B
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 1
Flash Point(F)
482.0 °F - closed cup
Flash Point(C)
250 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
c'est quoi la technique de dosage de l'Hexamethylenetetramine ?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service