Skip to Content
Merck
All Photos(1)

Key Documents

54265

Sigma-Aldrich

N-(2-Hydroxyethyl)aniline

technical, ≥98.0% (T)

Synonym(s):

2-(Phenylamino)ethanol, 2-Anilinoethanol, N-Phenylethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHCH2CH2OH
CAS Number:
Molecular Weight:
137.18
Beilstein:
774672
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)

grade

technical

Assay

≥98.0% (T)

refractive index

n20/D 1.578 (lit.)
n20/D 1.578

bp

278-282 °C/760 mmHg (lit.)

density

1.094 g/mL at 25 °C (lit.)

functional group

amine
hydroxyl

SMILES string

OCCNc1ccccc1

InChI

1S/C8H11NO/c10-7-6-9-8-4-2-1-3-5-8/h1-5,9-10H,6-7H2

InChI key

MWGATWIBSKHFMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - STOT RE 2 - STOT SE 1

Target Organs

Blood, Blood,hematopoietic system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mercedes Amat et al.
Natural product communications, 6(4), 515-526 (2011-05-13)
This review is focused on recent synthetic achievements and ongoing work in our laboratory using phenylglycinol-derived oxazolopiperidone lactams as starting materials for the enantioselective synthesis of piperidine-containing alkaloids: madangamines, 2,5-disubstituted decahydroquinoline and 1-substituted tetrahydroisoquinoline alkaloids, the indole alkaloids 20S- and
Mercedes Amat et al.
Organic & biomolecular chemistry, 9(7), 2175-2184 (2011-02-08)
The double cyclocondensation of symmetric pyridyl bis(oxoacids) 2b and 3b with (R)-phenylglycinol stereoselectively gave access to bis-phenylglycinol-derived oxazolopyrrolidine 9 and oxazolopiperidone 10, respectively. Application of the stereocontrolled cyclocondensation reaction to phenyl bis-γ-oxoacid 4b provided 11, which was converted to the
Mercedes Amat et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(28), 7724-7732 (2011-06-15)
Phenylglycinol-derived, unsaturated oxazolopiperidone lactams are extremely useful building blocks that undergo stereoselective conjugate addition reactions with organocuprates, enolates, and sulfur-stabilized anions, allowing the stereocontrolled introduction of substituents at the piperidine 4-position. The factors governing the exo- or endo-facial selectivity are
Valentine J Klimkowski et al.
Bioorganic & medicinal chemistry letters, 17(21), 5801-5805 (2007-09-21)
Analogs to a series of D-phenylglycinamide-derived factor Xa inhibitors were discovered. It was found that the S4 amide linkage can be replaced with an ether linkage to reduce the peptide character of the molecules and that this substitution leads to
Santos Fustero et al.
The Journal of organic chemistry, 74(11), 4429-4432 (2009-05-15)
The preparation of cyclic dipeptide mimetics from chiral imino lactones derived from (R)-phenylglycinol is described. Key steps of the synthetic route included the fully stereoselective construction of a quaternary center, the formation of six-, seven-, or eight-membered lactams by means

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service