Skip to Content
Merck
All Photos(1)

Key Documents

46818

Supelco

Sulfaguanidine monohydrate

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N4O2S · H2O
CAS Number:
Molecular Weight:
232.26
Beilstein:
5659891
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical
forensics and toxicology
pharmaceutical (small molecule)

SMILES string

O.NC(=N)NS(=O)(=O)c1ccc(N)cc1

InChI

1S/C17H13ClN2O2S/c18-13-5-1-12(2-6-13)10-22-14-7-3-11(4-8-14)9-15-16(21)20-17(19)23-15/h1-9H,10H2,(H2,19,20,21)/b15-9-

InChI key

LMMODKUVVDSJOZ-DHDCSXOGSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service