Skip to Content
Merck
All Photos(1)

Key Documents

38278

Supelco

DL-Menthol

reference material

Synonym(s):

Menthol, 2-Isopropyl-5-methylcyclohexanol, Hexahydrothymol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
Beilstein:
3194263
EC Number:
MDL number:
UNSPSC Code:
41116107

grade

reference material

Quality Level

vapor pressure

0.8 mmHg ( 20 °C)

Assay

≥98.0% (GC)

optical purity

enantiomeric ratio: ~50:50

shelf life

limited shelf life, expiry date on the label

bp

216 °C (lit.)

mp

34-36 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC(C)C1CCC(C)CC1O

InChI

1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3

InChI key

NOOLISFMXDJSKH-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.

DL-Menthol, a cyclic terpene alcohol and a naturally occurring compound, originates from the plants of Mentha species. It has a typical minty smell, flavor and finds applications as an ingredient in fragrances, food products, and cosmetics including shower gels, shampoos, face creams, body lotions, liqueurs, candy, etc.

Application

DL-Menthol may be used as an extraction solvent for the determination of parabens in food products, cosmetics and pharmaceuticals using liquid-liquid microextraction with high performance liquid chromatography-diode array detection (HPLC-DAD). It may also be used for the preparation of hydrophobic deep eutectic solvent (DES) in the extraction and determination of methylparaben, propylparaben, and butylparaben from cosmetics samples using syringe-to-syringe dispersive magnetic nanofluid microextraction procedure (SS-DMNF-ME) with high-performance liquid chromatography technique (HPLC).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

204.8 °F

Flash Point(C)

96 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of a New Polymeric Deep Eutectic Solvent and Its Application in Vortex-Assisted Liquid-Liquid Microextraction of Parabens in Foods, Cosmetics and Pharmaceutical Products
Ge D, et al.
Journal of the Brazilian Chemical Society, 31(10), 2120-2128 (2020)
Simultaneous selective enrichment of methylparaben, propylparaben, and butylparaben from cosmetics samples based on syringe-to-syringe magnetic fluid phase microextraction
Dil AE, et al.
Talanta, 221(8), 121547-121547 (2013)
Direct Determination of menthol using a simple spectrophotometric method
Anastasia-Sandu A, et al.
Bulletin of the Chemical Society of Japan, 46(8), 71-80 (2013)
R Eccles
The Journal of pharmacy and pharmacology, 46(8), 618-630 (1994-08-01)
Menthol and related cooling compounds such as 'coolant agent 10', are widely used in products ranging from common cold medications to toothpastes, confectionery, cosmetics and pesticides. The review brings together a range of information on production and chemistry of menthol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service