Skip to Content
Merck
All Photos(1)

Key Documents

37074

Supelco

Peroxide 1000 test sticks Quantofix®

50-1000 mg/L (H2O2)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319
NACRES:
NA.21
Pricing and availability is not currently available.

description

semi-quantitative

specific analyte(s)

hydrogen peroxide

reaction suitability

reaction type: Redox Reactions

measuring range

50-1000 mg/L (H2O2)

application(s)

environmental
food and beverages
general analytical
industrial hygiene

detection method

colorimetric

General description

Contents:
100 test sticks.

Storage:
Avoid exposing the strips to sunlight and moisture. Keep container cool and dry (storage temperature not above 30°C).

Gradation:
0 − 50 − 150 − 300 − 500 − 800 − 1000 mg/L H2O2

Application

Peroxide Quantofix® test sticks may be used to determine hydrogen peroxide concentration, in order to investigate if presence of H2O2 can selectively suppress cyanobacteria in lake without affecting biota.[1]

Legal Information

Quantofix is a registered trademark of Macherey-Nagel GmbH & Co., KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hans C P Matthijs et al.
Water research, 46(5), 1460-1472 (2011-11-25)
Although harmful cyanobacteria form a major threat to water quality, few methods exist for the rapid suppression of cyanobacterial blooms. Since laboratory studies indicated that cyanobacteria are more sensitive to hydrogen peroxide (H(2)O(2)) than eukaryotic phytoplankton, we tested the application

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service