Skip to Content
Merck
All Photos(1)

Key Documents

23170

Sigma-Aldrich

(2-Bromoethyl) chloroformate

purum, ≥95.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOOCH2CH2Br
CAS Number:
Molecular Weight:
187.42
Beilstein:
906849
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

grade

purum

Assay

≥95.0% (AT)

refractive index

n20/D 1.478 (lit.)
n20/D 1.478

bp

67-68 °C/11 mmHg

density

1.743 g/mL at 20 °C
1.749 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClC(=O)OCCBr

InChI

1S/C3H4BrClO2/c4-1-2-7-3(5)6/h1-2H2

InChI key

ZLCKDYMZBZNBMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

For the introduction of the BEOC-amino protecting group which is cleaved with diluted base after modification with tert-phosphines; Or with fluoride ions after modification with trimethylstannyl-lithium

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T.-L. Ho
Synthetic Communications, 8, 359-359 (1978)
H. Kunz
Justus Liebigs Annalen der Chemie, 1674-1674 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service