Skip to Content
Merck
All Photos(1)

Key Documents

19345

Supelco

Steviol

analytical standard

Synonym(s):

(4α)-13-Hydroxykaur-16-en-18-oic acid, ent-13-Hydroxykauran-16-en-19-oic acid, NSC 226902

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30O3
CAS Number:
Molecular Weight:
318.45
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water (Karl Fischer)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

C[C@@]12CCC[C@](C)([C@H]1CC[C@]34CC(=C)[C@](O)(CC[C@@H]23)C4)C(O)=O

InChI

1S/C20H30O3/c1-13-11-19-9-5-14-17(2,7-4-8-18(14,3)16(21)22)15(19)6-10-20(13,23)12-19/h14-15,23H,1,4-12H2,2-3H3,(H,21,22)/t14-,15-,17+,18+,19+,20-/m0/s1

InChI key

QFVOYBUQQBFCRH-VQSWZGCSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Steviol, a diterpenoiic carboxylic alcohol is an aglycone of sweet glycosides which is accumulated in Stevia rebaudiana. ent-kaurenoic acident-KA undergoes hydroxylation reaction in presence of molecular oxygen and NADPH, to yield steviol.[1][2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Steviol may have been used as reference standard to compare retention time and UV spectra in HPLC analysis for the quantification of steviol in Aconitum heterophyllum plant extract.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

K K Kim et al.
Archives of biochemistry and biophysics, 332(2), 223-230 (1996-08-15)
The diterpenoic compound steviol (ent-kaur-16-en-13-ol-19-oic acid) is the aglycone of sweet glycosides accumulated in Stevia rebaudiana Bertoni. This compound is the hydroxylated form of ent-kaurenoic acid (ent-kaur-16-en-19-oic acid; ent-KA). The hydroxylation of ent-KA to form steviol requiring NADPH and molecular
Claudio Gardana et al.
Journal of chromatography. A, 1217(9), 1463-1470 (2010-01-28)
Stevia rebaudiana leaves contain non-cariogenic and non-caloric sweeteners (steviol-glycosides) whose consumption could exert beneficial effects on human health. Steviol-glycosides are considered safe; nonetheless, studies on animals highlighted adverse effects attributed to the aglycone steviol. The aim of the present study
Stijn Ceunen et al.
Plant science : an international journal of experimental plant biology, 198, 72-82 (2012-12-04)
The effect of photoperiodism on steviol glycoside (SVgly) accumulation was investigated in Stevia rebaudiana. Topped plants were cultivated to develop new branches under a 16h or 8h photoperiod. During different ontogenetic phases, leaves, stems, lateral shoots, roots and reproductive organs
Molecular dissection of pathway components unravel atisine biosynthesis in a non-toxic Aconitum species, A. heterophyllum Wall.
Kumar, Varun, et al.
3 Biotech, 6.1, 1-10 (2016)
H Shibata et al.
Plant physiology, 95(1), 152-156 (1991-01-01)
To evaluate and characterize stevioside biosynthetic pathway in Stevia rebaudiana Bertoni cv Houten, two enzyme fractions that catalyze glucosylation of steviol (ent-13-hydroxy kaur-16-en-19-oic acid) and steviol-glucosides (steviol-13-O-glucopyranoside, steviolbioside and stevioside), utilizing UDP-glucose as the glucose donor, were prepared from the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service