Skip to Content
Merck
All Photos(1)

Key Documents

15837

Supelco

1-Methoxynaphthalene

analytical standard

Synonym(s):

Methyl 1-naphthyl ether, NSC 5530

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7OCH3
CAS Number:
Molecular Weight:
158.20
Beilstein:
774884
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.620-1.625
n20/D 1.621 (lit.)

bp

135-137 °C/12 mmHg (lit.)

density

1.09 g/mL at 25 °C (lit.)

format

neat

SMILES string

COc1cccc2ccccc12

InChI

1S/C11H10O/c1-12-11-8-4-6-9-5-2-3-7-10(9)11/h2-8H,1H3

InChI key

NQMUGNMMFTYOHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Methoxynaphthalene, a methoxyphenolic compound, is a naphthalene derivative that finds application as a precursor for the preparation of pharmaceutical drugs, and also as a therapeutic agent.

Application

1-Methoxynaphthalene can be used as an analytical standard for the determination of the analyte in tea beverages by gas chromatography-mass spectrometry (GC-MS) and high-performance liquid chromatography (HPLC) techniques.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oolong tea made from tea plants from different locations in Yunnan and Fujian, China showed similar aroma but different taste characteristics
Wang C, et al.
SpringerPlus, 5(1), 576-576 (2016)
Comparative analysis of Pu-erh and Fuzhuan teas by fully automatic headspace solid-phase microextraction coupled with gas chromatography-mass spectrometry and chemometric methods
Lv S, et al.
Journal of Agricultural and Food Chemistry, 62(8), 1810-1818 (2014)
Experimental (FT-IR and FT-Raman), electronic structure and DFT studies on 1-methoxynaphthalene
Govindarajan M, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 79(3), 646-653 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service