Skip to Content
Merck
All Photos(1)

Key Documents

15315

Supelco

Lead ion standard solution

for ion-selective electrodes

Synonym(s):

Lead(II) nitrate solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
26111700

grade

for ion-selective electrodes

shelf life

limited shelf life, expiry date on the label

concentration

0.1 M Pb

density

1.00 g/mL at 20 °C

SMILES string

[Pb]

InChI

1S/Pb

InChI key

WABPQHHGFIMREM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Preparation Note

prepared with Pb(NO3)2 and H2O

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service