Assay
≥97.0% (CHN)
mp
65-68 °C (lit.)
66-68 °C
storage temp.
2-8°C
SMILES string
[N-]=[N+]=Nc1ccc(cc1)N=C=S
InChI
1S/C7H4N4S/c8-11-10-7-3-1-6(2-4-7)9-5-12/h1-4H
InChI key
XMPGXADJZXSISQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
Heterobifunctional crosslinking agent which forms a crosslink over a range of 10 Å and is expected to facilitate the investigation of hydrophobically interacting sites between proteins and membrane lipids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Methods in Enzymology, 88, 207-207 (1982)
Febs Letters, 113, 307-307 (1980)
International journal of pharmaceutics, 404(1-2), 205-210 (2010-11-09)
Staudinger ligation was evaluated as a strategy for synthesizing receptor targeted liposomes. First, an activated lipid derivative was synthesized by reacting dioleoyl phosphatidylethanolamine (DOPE) and 2-(diphenylphosphino) terephthalic acid 1-methyl 4-penta-fluorophenyldiester. Second, transferrin (Tf) was activated with p-azidophenyl isothiocyanate. Third, liposomes
European journal of biochemistry, 125(1), 197-201 (1982-06-15)
Two hydrophobic, heterobifunctional azidoarylisothiocyanates have been synthesized. The reagents are applicable for group-directed modification of membrane-integrated protein segments and subsequent photo-induced cross-linking with neighbouring membrane constituents. Both 5-isothiocyanato-1-naphthalene azide and p-azidophenylisothiocyanate are chemically characterized with respect to structure and photosensitivity.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service