Skip to Content
Merck
All Photos(1)

Key Documents

11332

Sigma-Aldrich

Atropine sulfate salt monohydrate

tested according to Ph. Eur.

Synonym(s):

Atropini sulfas, α-(Hydroxymethyl)benzeneacetic acid 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, Tropine tropate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C17H23NO3)2 · H2SO4 · H2O
CAS Number:
Molecular Weight:
694.83
Beilstein:
6109275
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
tested according to Ph. Eur.

mp

189-192 °C (A) (lit.)

solubility

water: soluble 2500 g/L at 4 °C

application(s)

pharmaceutical (small molecule)

SMILES string

O.OS(O)(=O)=O.CN1[C@H]2CC[C@@H]1C[C@@H](C2)OC(=O)C(CO)c3ccccc3.CN4[C@H]5CC[C@@H]4C[C@@H](C5)OC(=O)C(CO)c6ccccc6

InChI

1S/2C17H23NO3.H2O4S.H2O/c2*1-18-13-7-8-14(18)10-15(9-13)21-17(20)16(11-19)12-5-3-2-4-6-12;1-5(2,3)4;/h2*2-6,13-16,19H,7-11H2,1H3;(H2,1,2,3,4);1H2/t2*13-,14+,15+,16?;;

InChI key

JPKKQJKQTPNWTR-CHYDPLAESA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Atropine sulfate salt monohydrate has been used for anaesthetizing mice for intranasal administration of ropinirole or 7-OH-DPAT.

Biochem/physiol Actions

Atropine sulfate salt monohydrate is a competitive nonselective antagonist of central and peripheral muscarinic acetylcholine receptors. Atropine is a cholinergic receptor antagonist isolated from Atropa belladona L., Datura stramonium L. and other plants of the solanaceae family. It blocks the ion currents mediated by neuronal nicotinic acetylcholine receptors. Further, it blocks the acetylcholine (ACh)-induced relaxations in human pulmonary veins.
Competitive nonselective antagonist at central and peripheral muscarinic acetylcholine receptors.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Trovero et al.
Toxicology and applied pharmacology, 150(2), 321-327 (1998-07-07)
This study consists of two parts, first to compare the pharmacological profile of atropine and CEB-1957 substance toward muscarinic receptor subtypes. In various rat brain structures, binding properties were determined by competition experiments of [3H]pirenzepine, [3H]AF-DX 384, and [3H]4-DAMP in
L Walch et al.
British journal of pharmacology, 130(1), 73-78 (2000-04-26)
1. To characterize the muscarinic receptors on human pulmonary veins associated with the acetylcholine (ACh)-induced relaxation, isolated venous and arterial preparations were pre-contracted with noradrenaline (10 microM) and were subsequently challenged with ACh in the absence or presence of selective
Chu Lan Lao et al.
Neurotoxicity research, 24(4), 523-531 (2013-07-04)
Parkinson's disease (PD) is a neurodegenerative disease with a hallmark motor defect caused by the death of dopaminergic neurons in the substantia nigra. Intranasal drug administration may be useful for Parkinson's treatment because this route avoids first-pass metabolism and increases
R Zwart et al.
Molecular pharmacology, 52(5), 886-895 (1997-11-14)
Atropine, the classic muscarinic receptor antagonist, inhibits ion currents mediated by neuronal nicotinic acetylcholine receptors expressed in Xenopus laevis oocytes. At the holding potential of -80 mV, 1 microM atropine inhibits 1 mM acetylcholine-induced inward currents mediated by rat alpha2beta2
Miklós Palotai et al.
Neurochemical research, 39(8), 1477-1483 (2014-06-05)
The nociceptin/orphanin FQ peptide (NOP) receptor and its endogenous ligand plays role in several physiologic functions of the central nervous system, including pain, locomotion, anxiety and depression, reward and drug addiction, learning and memory. Previous studies demonstrated that the NOP-receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service