Skip to Content
Merck
All Photos(1)

Key Documents

06712

Supelco

3,4,5-Trimethoxycinnamic acid

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3C6H2CH=CHCO2H
CAS Number:
Molecular Weight:
238.24
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

125-127 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

COc1cc(\C=C\C(O)=O)cc(OC)c1OC

InChI

1S/C12H14O5/c1-15-9-6-8(4-5-11(13)14)7-10(16-2)12(9)17-3/h4-7H,1-3H3,(H,13,14)/b5-4+

InChI key

YTFVRYKNXDADBI-SNAWJCMRSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[The detection of Azotobacter and its significance in criminal technical soil examinations].
E Kissling
Archiv fur Kriminologie, 165(1-2), 27-34 (1980-01-01)
Sarvesh Kumar et al.
Biochemistry, 44(48), 15944-15952 (2005-11-30)
We report here the isolation and characterization of two active principles, ethyl 3',4',5'-trimethoxycinnamate (1) and piperine (2), from the combined hexane and chloroform extracts of Piper longum. Using primary human umbilical vein endothelial cells, we evaluated the activities of compound
Zhenghang Zhao et al.
Biological & pharmaceutical bulletin, 36(2), 238-244 (2012-12-01)
3,4,5-Trimethoxycinnamic acid (TMCA), methyl 3,4,5-trimethoxycinnamate (M-TMCA) and p-methoxycinnamic acid (PMCA) have been identified as the major bioactive components in the serum collected from rats treated with oral administration of Polygalae Radix ("YuanZhi," the roots of Polygala tenuifolia WILLD.), a traditional
Keiko Kawashima et al.
Biological & pharmaceutical bulletin, 27(8), 1317-1319 (2004-08-12)
3,4,5-trimethoxycinnamic acid (TMCA) is one of the constituents in Onji (roots of Polygala tenuifolia WILLD), an herbal medicine used for sedative in Japanese traditional Kampo medicine. Our previous study revealed that oral administration of this compound prolongs sleeping time induced
M I Donnelly et al.
Journal of bacteriology, 147(2), 471-476 (1981-08-01)
When grown on 3,4,5-trimethoxycinnamic acid, a strain of Pseudomonas putida oxidized this compound and also 3,4,5-trimethoxybenzoic, 3,5-dimethoxy-4-hydroxybenzoic (syringic), and 3,4-dihydroxy-5-methoxybenzoic (3-O-methylgallic) acids, but 3,5-dimethoxy-4-hydroxycinnamic and other acids bearing structural resemblances to the growth substrate were oxidized only slowly. These results

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service