Skip to Content
Merck
All Photos(1)

Key Documents

00488

Sigma-Aldrich

Helium

≥99.999%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
He
CAS Number:
Molecular Weight:
4.00
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

0.14 (vs air)

Quality Level

Assay

≥99.999%

bp

-268.934 °C (lit.)
−268.9 °C (lit.)

mp

-272.2 °C (lit.)

SMILES string

[He]

InChI

1S/He

InChI key

SWQJXJOGLNCZEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Pressure tin
Filling pressure at 15°C: 11 Bar; content 15°C, 1 bar): 11 l

Other Notes

Sales restrictions may apply

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service