Skip to Content
Merck
All Photos(1)

Key Documents

MABF2113

Sigma-Aldrich

Anti-EBOV GP Antibody, clone 1H3

clone 1H3, from mouse

Synonym(s):

Envelope glycoprotein, GP1, 2, GP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1H3, monoclonal

species reactivity

virus, Ebola virus

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
neutralization: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

General description

Envelope glycoprotein (UniProt: Q05320; also known as GP1,2; GP) is encoded by the GP gene (Gene ID: 911829) in Zaire Ebola virus. Ebola virus (EBOV) is a filovirus that is shown to cause severe viral hemorrhagic fever with high lethality. It has a single stranded, negative-sense RNA genome that encodes seven viral structural proteins including nucleoprotein, virion proteins, and glycoprotein. The glycoprotein is the most important protein involved in pathogenesis. It is a type I transmembrane surface protein shown to be responsible for receptor binding, viral entry, and cellular tropism. The GP gene is reported to undergo transcriptional editing to give rise to several glycosylated proteins, including the structural protein GP1,2, and the secreted non-structural glycoprotein (sGP). The GP is synthesized as a 676-amino acid polyprotein with a signal peptide (aa 1-32). This polyprotein is cleaved by the furin to yield two disulfide linked subunits known as GP1 (aa 33-501) and GP2 (aa 502-676), which together form the GP1,2 heterodimer. The GP1 acts as the receptor-binding subunit and GP2 as the membrane fusion subunit. The GP1 subunit is expressed on the cell surface with the C-terminus oriented toward the aqueous environment, while the N-terminus is bound to GP2 via disulfide bonds. The GP1 subunit contains the heavily glycosylated mucin-like domain (aa 305-485), which is responsible for its cytotoxic function. The glycoprotein contains two coiled coil regions (aa 554-595 and 615-634) that play a role in its oligomerization and fusion activity. Clone 1H3 recognizes the secreted glycoprotein form (sGP) and also recognizes glycoprotein lacking a large part of the mucin domain (GP1,2 Mucin333 458). It does not recognize VSV G/ZEBOVGP and subfragment D. When administered before virus challenge, it provides only a partial protection. (Ref.: Qiu, X., et al. (2011). Clin. Immunol. 141(2); 218-227).

Specificity

Clone 1H3 is a mouse monoclonal antibody that detects envelope glycoprotein of Zaire Ebola virus. It recognizes the secreted glycoprotein form (sGP) and binds to a conformational epitope in the first 295 amino acids.

Immunogen

Glycoprotein from Zaire Ebola virus, strain Mayinga, expressed in VSV G. Reverse genetics technique was used to create the recombinant virus VSV G/ZEBOV glycoprotein.

Application

Anti-EBOV GP clone 1H3, Cat. No. MABF2113, is a mouse monoclonal antibody that detects Ebola virus Envelope Glycoprotein and has been tested for use in ELISA and, Immunocytochemistry, Immunoprecipitation, and Neutralizing Applications.
ELISA Analysis: A representative lot detected EBOV GP in ELISA applications (Qiu, X., et. al. (2011). Clin Immunol. 141(2):218-27).

Immunoprecipitation Analysis: Immunoprecipitation Analysis: A representative lot detected EBOV GP in Immunoprecipitation applications (Qiu, X., et. al. (2011). Clin Immunol. 141(2):218-27).

Immunocytochemistry Analysis: Immunocytochemistry Analysis: A representative lot detected EBOV GP in Immunocytochemistry applications (Qiu, X., et. al. (2011). Clin Immunol. 141(2):218-27).

Neutralizing Analysis: A representative provided partial protection when administered prior to virus challenge. (Qiu, X., et. al. (2011). Clin Immunol. 141(2):218-27).

ELISA Analysis: Various dilutions of this antibody detected Envelope glycoprotein in Recombinant Ebola virus Glycoprotein minus the transmembrane region (EBOV rGP TM).
Research Category
Inflammation & Immunology

Quality

Evaluated by ELISA with recombinant Ebola virus Glycoprotein minus the transmembrane region (EBOV rGP deltaTM).

ELISA Analysis: Various dilutions of this antibody detected EBOV GP in Recombinant Ebola virus Glycoprotein minus the transmembrane region (EBOV rGP deltaTM).

Target description

74.46 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service