Skip to Content
Merck
All Photos(1)

Documents

MAB947

Sigma-Aldrich

Anti-Coxsackievirus A9 Antibody, clone 450-11G-8A-5E

ascites fluid, clone 450-11G-8A-5E, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

450-11G-8A-5E, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG2b

shipped in

wet ice

Specificity

Reacts with the Coxsackie virus A9. Neutralizes coxsackie virus A9 Griggs strain at a titer of 1:112.

Application

Anti-Coxsackievirus A9 Antibody, clone 450-11G-8A-5E is an antibody against Coxsackievirus A9 for use in IF.
IFA at 1:1,000. Dilute with buffer pH 7.5-8.0 containing protein or other stabilizing medium. Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Ascites

Storage and Stability

Maintain at -20°C for up to 12 months in convenient aliquots. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integrin alpha v beta 6 is an RGD-dependent receptor for coxsackievirus A9.
Williams, CH; Kajander, T; Hyypia, T; Jackson, T; Sheppard, D; Stanway, G
Journal of virology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service