Skip to Content
Merck
All Photos(1)

Key Documents

8.03138

Sigma-Aldrich

Isopropenyl acetate

for synthesis

Synonym(s):

Isopropenyl acetate, Acetone enol acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC(CH3)=CH2
CAS Number:
Molecular Weight:
100.12
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-562-7
Pricing and availability is not currently available.

vapor pressure

23 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

439 °C (DIN 51794)

potency

3000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

expl. lim.

1.8 % (v/v)

pH

3 (20 °C, 34 g/L in H2O)

bp

97 °C/1013 hPa

mp

-93 °C

transition temp

flash point 16 °C

density

0.92 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C5H8O2/c1-4(2)7-5(3)6/h1H2,2-3H3

InChI key

HETCEOQFVDFGSY-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.920 - 0.923
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

66.2 °F - closed cup

Flash Point(C)

19 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service