Skip to Content
Merck
All Photos(1)

Key Documents

8.01516

Sigma-Aldrich

Benzylideneacetone

for synthesis

Synonym(s):

Benzylideneacetone, 4-Phenyl-3-buten-2-one, Methyl styryl ketone, Benzalacetone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O
CAS Number:
Molecular Weight:
146.19
MDL number:
UNSPSC Code:
12352115
EC Index Number:
204-555-1

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

potency

2031 mg/kg LD50, oral (Rat)

bp

261 °C/1013 hPa

mp

38-41 °C

transition temp

flash point 116 °C (DIN 51758)

solubility

1.3 g/L

density

1.008 g/cm3 at 47 °C

storage temp.

2-25°C

InChI

1S/C10H10O/c1-9(11)7-8-10-5-3-2-4-6-10/h2-8H,1H3/b8-7+

InChI key

BWHOZHOGCMHOBV-BQYQJAHWSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 38 °C
Melting range (upper value): ≤ 41 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

253.4 °F - closed cup

Flash Point(C)

123 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service